Please login to your online account to display your discounted pricing

Potassium Chloride Conductivity Standard, 5 μS/cm (5 0μmho/cm) at 25°C, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Chloride Conductivity Standard
Name Note 5μS/cm (50μmho/cm) at 25°C
CAS 7447-40-7
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 50.0 ± 0.1 μS/cm at 25°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R588650010F

 
ricca chemical company
RIR588650010F
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R588650020F

 
ricca chemical company
RIR588650020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 10L
Name Note 5μS/cm (50μmho/cm) at 25°C
Solvent or Matrix Aqueous Solution
Packaging Cubitainer
CAS 7447-40-7
Assay Percent Range 50.0 ± 0.1 μS/cm at 25°C
Solubility Solubility in Water: Inifinite
CAS 7732-18-5
Odor Odorless
Chemical Name or Material Potassium Chloride Conductivity Standard