Please login to your online account to display your discounted pricing

Potassium Chloride Conductivity Standard, 2500μS/cm at 25°C, Ricca Chemical


Quantity 10L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR588802010F

 View more versions of this product

Catalog No. R588802010F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 10L
Chemical Name or Material Potassium Chloride Conductivity Standard
CAS 7447-40-7
Name Note 2500μS/cm at 25°C
Assay Percent Range 2, 500 ± 10 μS/cm at 25°C
Odor Odorless
CAS 7732-18-5
Packaging Cubitainer
Solubility Solubility in Water: Infinite

Specifications

Chemical Name or Material Potassium Chloride Conductivity Standard
Name Note 2500μS/cm at 25°C
CAS 7447-40-7
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 2, 500 ± 10 μS/cm at 25°C