missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Chloride Conductivity Standard, 2500 μS/cm at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company R588802020F
Specifications
Potassium Chloride Conductivity Standard | |
7447-40-7,7732-18-5 | |
20 L | |
2, 500 ± 10 μS/cm at 25°C | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
Odorless | |
Solubility in Water: Infinite |
2500μS/cm at 25°C | |
Cubitainer | |
Colorless | |
ClK | |
[Cl-].[K+] | |
74.548 | |
CHEBI:32588 | |
Aqueous Solution |