missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Chloride Conductivity Standard, 1000 μS/cm at 25°C, Ricca Chemical
Tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.
Supplier: Ricca Chemical Company 58880132
Description
- Tested in ISO 17025 Accredited Laboratories.
- Manufactured in cGMP compliant, FDA registered facilities.
- Certified Traceable to NIST Standard Reference Material when available.
- Many C of As and labels contain TDS and conductance information.
- Tightest Specifications in the industry, lowest lot-to-lot variability.
Specifications
Potassium Chloride Conductivity Standard | |
7447-40-7,7732-18-5 | |
1 L | |
100.0%,0.05% | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
Liquid |
1 mS/cm (1000μmho/cm) at 25°C | |
Natural Poly Bottle | |
98.0%,0.05% | |
Colorless | |
MFCD00011360 | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 |
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
4873 | |
potassium chloride |