Please login to your online account to display your discounted pricing

Potassium Chloride, ACS Grade, 99.0 to 100.5%, LabChem™

$73.66 - $73.66

Chemical Identifiers

Chemical Name or Material Potassium Chloride
CAS 7447-40-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187901

 
lab chem
LC187901
500g Each for $73.66
Description & Specifications

Specifications

Formula Weight 74.55
Chemical Name or Material Potassium Chloride
EINECS Number 231-211-8
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Assay 99.0 to 100.5%
Solubility Soluble in water
Specific Gravity 1.987g/cm3
Storage Information Room Temperature
Linear Formula KCl
CAS 7447-40-7
Decomposition Information Hydrogen chloride; Potassium oxide
Boiling Point 1,420°C
Color White
Melting Point 770°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Density 1.987g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.