Please login to your online account to display your discounted pricing

Potassium Chloride ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  685806

 View more versions of this product

Catalog No. MK6858212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 74.55
Purity Grade ACS Grade
Quantity 2.5kg
Packaging Poly
Molecular Formula KCl
Physical Form Granular
Chemical Name or Material Potassium Chloride
CAS 7447-40-7

Specifications

Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula KCl
Formula Weight 74.55