missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Chloride ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7447-40-7
Physical Form Granular
Molecular Formula KCl
Formula Weight 74.55
Chemical Name or Material Potassium Chloride
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK6858212 Avantor Performance
685806
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK6858500 Avantor Performance
685804
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7447-40-7
KCl
Potassium Chloride
Granular
74.55
ACS Grade

Specifications

7447-40-7
KCl
Poly
Potassium Chloride
Granular
74.55
2.5kg
ACS Grade