Please login to your online account to display your discounted pricing

Potassium Chloride ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging Flowmor

Manufacturer: avantor performance  685820

 View more versions of this product

Catalog No. MK685812

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Flowmor
Quantity 12kg
Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Purity Grade ACS
Physical Form Granular
Molecular Formula KCl
Formula Weight 74.55g/mol

Specifications

Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Purity Grade ACS
Physical Form Granular
Molecular Formula KCl
Formula Weight 74.55g/mol