missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium chloride, ACS, 99.0-100.5%, Alfa Aesar™

In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy

Manufacturer:  Alfa Aesar 1159530

 View more versions of this product

Catalog No. AA1159530



Description

Description

  • In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy
Specifications

Specifications

Potassium chloride
7447-40-7
ClK
14,7621
Freely soluble in water; Soluble in glycerol, alcohol
[Cl-].[K+]
74.548
CHEBI:32588
Crystalline
ACS
Odorless
Hygroscopic
773°C
For analysis ACS
99.0 to 100.5%
MFCD00011360
chlorvescent
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
99.0 to 100.5%
1.984g/mL
1.490
1500°C (sublimation)
250g

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
chlorvescent
CHEBI:32588
[Cl-].[K+]
Safety and Handling

Safety and Handling

RTECSNumber : TS8050000

TSCA : TSCA

Documents