Please login to your online account to display your discounted pricing

Potassium chloride, 99+%, for analysis, ACROS Organics™

$173.74 - $173.74

Chemical Identifiers

Chemical Name or Material Potassium chloride
Name Note p.a.
CAS 7447-40-7
Purity Grade For analysis
Assay 99.0 to 100.5% (calc. on dried substance) (Argentometry)
Assay Percent Range 99+%
Molecular Formula ClK
Linear Formula KCl
Formula Weight 74.54
MDL Number MFCD00011360
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC196770050

  SDS
thermo fisher scientific
AC196770050
Plastic bottle 5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC196770010

  SDS
fisher scientific acros
196770010
Plastic bottle 1kg Each for $173.74

AC196770250

  SDS
thermo fisher scientific
AC196770250
Fiber drum 25kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 99.0 to 100.5% (calc. on dried substance) (Argentometry)
Physical Form Liquid
Formula Weight 74.54
Quantity 5kg
Color Clear colorless
TSCA TSCA
RTECS Number TS8050000
EINECS Number 231-211-8
Purity Grade For analysis
Sublimation Point 1500.0°C
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Calcium (Ca) 50ppm max.
Magnesium (Mg) 50ppm max.
Trace Analysis 20ppm max.
Cadmium (Cd) 1ppm max.
Name Note p.a.
Chemical Name or Material Potassium chloride
MDL Number MFCD00011360
Linear Formula KCl
Assay Percent Range 99+%
CAS 7447-40-7
Molecular Formula ClK
Iron (Fe) 10ppm max.
Packaging Plastic bottle
Sulfate 100ppm max.
Heavy Metals (as Pb) 5ppm max.
Aluminum (Al) 1ppm max.
Iodide 50ppm max.
Bromide 1000ppm max.
Lead (Pb) 2ppm max.
Copper (Cu) 10ppm max.
Sodium (Na) 700ppm max.
Arsenic (As) 0.5ppm max.
Acidity Alkalinity complies
Free Acid 0.003% max.
Bromine (Br) 1000ppm max.
Free Alkali 0.005% max.
Health Hazard 1 HYGROSCOPIC

Biological buffer for analysis

Safety and Handling

HYGROSCOPIC