Please login to your online account to display your discounted pricing

Potassium chloride, 99.997% (metals basis), Alfa Aesar™, Puratronic™

$171.77 - $721.74

Chemical Identifiers

Chemical Name or Material Potassium chloride
CAS 7447-40-7
Physical Form Powder/Crystalline/Beads
Assay Percent Range 99.997% (metals basis)
Molecular Formula KCl
Formula Weight 74.55
MDL Number MFCD00011360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1083930

  SDS
alfa aesar
1083930
250g Each for $721.74

AA1083918

  SDS
alfa aesar
1083918
50g Each for $171.77
Description & Specifications

Specifications

Physical Form Powder/Crystalline/Beads
Formula Weight 74.55
Quantity 250g
Chemical Name or Material Potassium Chloride
Boiling Point 1500°C (2732°F)
Melting Point 773°C (1423°F)
Color White
Density 1.984g/cm3 at 20drg.C
EINECS Number 231-211-8
Odor Odorless
Assay Percent Range 99.997% (metals basis)
Molecular Formula KCl
Appearance White Powder/crystalline/beads
MDL Number MFCD00011360
CAS 7447-40-7
Health Hazard 1 H303: May be harmful if swallowed.
Safety and Handling

H303: May be harmful if swallowed.