Learn More
Potassium chloride, 99.995% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 08762630
Description
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1500°C (sublimation) | |
773°C | |
White | |
250 g | |
ClK | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
74.55 | |
1.49 | |
1.984 |
Potassium chloride | |
7447-40-7 | |
Powder | |
99.995% (metals basis) | |
MFCD00011360 | |
Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether. | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
Odorless | |
Hygroscopic |
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 |
Safety and Handling
H303: May be harmful if swallowed.
EINECSNumber : 231-211-8
RTECSNumber : TS8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only