missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride, 99%

Manufacturer:  Alfa Aesar™ A116620B

 View more versions of this product

Catalog No. AAA116620B



Description

Description

Applications
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

Solubility
Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether.

Notes
Hygroscopic.
Specifications

Specifications

Potassium chloride
99%
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
1500°C (sublimation)
1000g
7447-40-7
ClK
14,7621
[Cl-].[K+]
74.548
CHEBI:32588
Odorless
773°C

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium;chloride
Safety and Handling

Safety and Handling

RTECSNumber : TS8050000

TSCA : TSCA

SDS Documents