Please login to your online account to display your discounted pricing

Potassium chloride, 99.0 to 100.5%, ACS, EMD Millipore


Quantity 500g

Manufacturer: emd millipore corporation  PX14051

 View more versions of this product

Catalog No. MPX14051

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS/EP/ISO/Reagent Grade
Physical Form Crystals
Formula Weight 74.55
Color White
Quantity 500g
Bulk Density 1000kg/m3
Density 1.98g/cm3 (20°C)
Chemical Name or Material Potassium chloride
Name Note EMSURE™, For Analysis
Assay Percent Range 99.0 to 100.5%
Molecular Formula ClK
Linear Formula KCl
CAS 7447-40-7
Recommended Storage Store without restrictions

Specifications

Chemical Name or Material Potassium chloride
Name Note EMSURE™, For Analysis
CAS 7447-40-7
Purity Grade ACS/EP/ISO/Reagent Grade
Physical Form Crystals
Assay Percent Range 99.0 to 100.5%
Molecular Formula ClK
Linear Formula KCl
Formula Weight 74.55
Accessories

Store without restrictions