Please login to your online account to display your discounted pricing

Potassium chloride, 99.0 to 100.5%, ACS, EMD Millipore

$110.23 - $508.26

Chemical Identifiers

Chemical Name or Material Potassium chloride
Name Note EMSURE™, For Analysis
CAS 7447-40-7
Purity Grade ACS/EP/ISO/Reagent Grade
Physical Form Crystals
Assay Percent Range 99.0 to 100.5%
Molecular Formula ClK
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX14051

 
emd millipore corporation
PX14051
500g Each for $110.23

MPX14057

 
emd millipore corporation
PX14057
12kg Each for $508.26

MPX14055

 
emd millipore corporation
PX14055
2.5kg Each for $329.71
Description & Specifications

Specifications

Physical Form Crystals
Purity Grade ACS/EP/ISO/Reagent Grade
Formula Weight 74.55
Color White
Quantity 500g
Bulk Density 1000kg/m3
Density 1.98g/cm3 (20°C)
Linear Formula KCl
Recommended Storage Store without restrictions
Molecular Formula ClK
Chemical Name or Material Potassium chloride
CAS 7447-40-7
Assay Percent Range 99.0 to 100.5%
Name Note EMSURE™, For Analysis
Safety and Handling

Store without restrictions