Please login to your online account to display your discounted pricing

Potassium Chloride, 3 Molar, saturated with Silver Chloride, Electrode Filling Solution, Ricca Chemical


Quantity 120mL

Manufacturer: ricca chemical company  RIR5918000120A

 View more versions of this product

Catalog No. R5918000120

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Potassium Chloride
Name Note Electrode Filling Solution
CAS 7732-18-5
CAS Min % 80.0
CAS Max % 83.0
CAS 7447-40-7
CAS Min % 17.89
CAS Max % 18.62
CAS 7761-88-8
CAS Min % 0.0034
CAS Max % 0.0036
Physical Form ear colorless liquid
Quantity 120mL

Specifications

Chemical Name or Material Potassium Chloride
Name Note Electrode Filling Solution
CAS 7732-18-5
CAS Min % 80.0
CAS Max % 83.0
CAS 7447-40-7
CAS Min % 17.89
CAS Max % 18.62
CAS 7761-88-8
CAS Min % 0.0034
CAS Max % 0.0036
Physical Form ear colorless liquid