Please login to your online account to display your discounted pricing

Potassium Chloride, 3.0M (3.0N), LabChem™

$33.17 - $33.17

Chemical Identifiers

Chemical Name or Material Potassium Chloride, 3.0M (3.0N)
CAS 7732-18-5
CAS Max % 80
CAS 7447-40-7
CAS Max % 20
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187951

 
lab chem
LC187951
500mL Each for $33.17
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 74.55
Chemical Name or Material Potassium Chloride, 3.0M (3.0N)
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1.13g/mL
Storage Information Room Temperature
Linear Formula KCl
CAS 7732-18-5
Viscosity 0.893cSt
Decomposition Information Hydrogen chloride; Potassium oxide
Color clear, colorless solution
Quantity 500mL
Traceability to NIST Traceable to NIST
CAS Max % 80
CAS 7447-40-7
CAS Max % 20
Concentration 3.00M ± 0.05M
Density 1.13g/mL