missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride, 1M aq. soln., RNAse free

$49.04 - $78.03

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.548
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62422EQE View Documents Alfa Aesar™
J62422EQE
125mL Each for $49.04
 
AAJ62422AK View Documents Alfa Aesar™
J62422AK
250mL Each for $78.03
 
Description

Description

Applications
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)

Solubility
Not miscible in water.

Notes
Store at room temperature. Ensure good ventilation.
Specifications

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]

Specifications

7447-40-7
KCl
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.548
CHEBI:32588
Liquid
125mL
ClK
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
1M aq. soln.
Potassium chloride, RNAse free
Safety and Handling

Safety and Handling

TSCA : TSCA