missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium chloride, 0.159M, Conductivity Standard, Thermo Scientific Chemicals
$58.04 - $194.40
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42697AP
|
Thermo Scientific Chemicals
042697AP |
500 mL |
Each for $58.04
|
|
|||||
AA42697K2
|
Thermo Scientific Chemicals
042697K2 |
1 L |
Each for $79.92
|
|
|||||
AA42697K7
|
Thermo Scientific Chemicals
042697K7 |
4 L |
Each for $194.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 |
ClK | |
MFCD00011360 | |
4873 | |
[Cl-].[K+] |
Specifications
7447-40-7,7732-18-5 | |
Colorless | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 | |
74.55 | |
23 hPa (17mm Hg) at 20°C | |
Potassium chloride, 20, 000μmho at 25°C |
500 mL | |
0.159M | |
MFCD00011360 | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
Potassium chloride: 1.2%; Water: 98.8% | |
Liquid |
Safety and Handling
EINECSNumber : 231-211-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only