missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride, 0.00017M, Conductivity Standard

Manufacturer:  Alfa Aesar™ 43444K7

 View more versions of this product

Catalog No. AA43444K7



Specifications

Specifications

Potassium chloride Conductivity Standard
7447-40-7
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
Odorless
25μmho at 25°C
ClK
0.00017M
[Cl-].[K+]
74.548
CHEBI:32588
Liquid
4L

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium;chloride
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents