missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium chloride,≈0.000027M, Conductivity Standard

Manufacturer:  Alfa Aesar™ 44065AP

 View more versions of this product

Catalog No. AA44065AP



Specifications

Specifications

Potassium chloride Conductivity Standard
7447-40-7
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
Odorless
10μmho at 25°C
ClK
≈0.000027M
[Cl-].[K+]
74.548
CHEBI:32588
Liquid
500mL

Chemical Identifiers

7447-40-7
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium;chloride
Safety and Handling

Safety and Handling

EINECSNumber : 231-211-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents