Learn More
Potassium carbonate sesquihydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
For Manufacturing of Soaps, Pigments, Printing Ink
Supplier: Thermo Scientific Chemicals 01098122
Description
Potassium carbonate sesquihydrate is used for manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts and as an analytical lab reagent. It is also useful as a reagent for removing carbon dioxide from ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent
Specifications
Potassium carbonate sesquihydrate | |
Crystalline | |
98.5-101.0% | |
MFCD00149925 | |
Soluble in water; Practically insoluble in alcohol | |
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O | |
330.45 | |
98.5 to 101.0% | |
Odorless |
6381-79-9 | |
100 g | |
C2H6K4O9 | |
14,7670 | |
WWVGGBSGYNQKCY-UHFFFAOYSA-J | |
tetrapotassium trihydrate dicarbonate | |
165.23 (138.21 Anhydrous) | |
ACS | |
Hygroscopic |
Chemical Identifiers
6381-79-9 | |
330.45 | |
WWVGGBSGYNQKCY-UHFFFAOYSA-J | |
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O |
C2H6K4O9 | |
MFCD00149925 | |
tetrapotassium trihydrate dicarbonate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-529-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only