Learn More
Potassium carbonate sesquihydrate, 98.5%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 347202500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium carbonate sesquihydrate | |
For analysis ACS | |
6381-79-9 | |
250 g | |
C2H6K4O9 | |
15, 7740 | |
WWVGGBSGYNQKCY-UHFFFAOYSA-J | |
tetrapotassium trihydrate dicarbonate | |
165.24 | |
ACS Reagent |
891.0°C | |
White | |
Crystalline Powder | |
98.5% | |
MFCD00149925 | |
Solubility in water: soluble in water., insoluble in alcohol | |
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O | |
330.45 | |
98.5% | |
Glass bottle |
Chemical Identifiers
6381-79-9 | |
330.45 | |
WWVGGBSGYNQKCY-UHFFFAOYSA-J | |
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O |
C2H6K4O9 | |
MFCD00149925 | |
tetrapotassium trihydrate dicarbonate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only