Please login to your online account to display your discounted pricing

Potassium Carbonate, Reagent ACS, Anhydrous, +99%, ACROS Organics™


Quantity 10kg
Packaging Plastic Drum

Manufacturer: thermo fisher scientific  AC424080100

 View more versions of this product

Catalog No. AC424080100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Granular Powder
Assay ≥99.0%
Purity Grade ACS Reagent Grade
Formula Weight 138.21
Quantity 10kg
Packaging Plastic Drum
Chemical Name or Material Potassium Carbonate
Melting Point 891°C
Color White
Merck Index 15, 7740
TSCA TSCA
EINECS Number 209-529-3
RTECS Number TS7750000
Fieser 01,1014; 04,50; 05,552; 08,408; 09,382; 10,323; 11,433; 12,403; 13,233; 15,167
Iron (Fe) 5ppm max.
Calcium (Ca) 50ppm max.
Heavy Metals (as Pb) 5ppm max.
Sulfur Compounds 0.004% max.
Insoluble Matter 0.01% max.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Assay Percent Range 99+%
Linear Formula K2CO3
Molecular Formula CK2O3
MDL Number MFCD00011382
CAS 584-08-7
Phosphate 10ppm max.
Chlorine (Cl) 30ppm max.
Sodium (Na) 200ppm max.
Magnesium (Mg) 20ppm max.
Silicon Dioxide 50ppm max.
Health Hazard 1 Warning

Specifications

Chemical Name or Material Potassium Carbonate
CAS 584-08-7
Purity Grade ACS Reagent Grade
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula CK2O3
Linear Formula K2CO3
Formula Weight 138.21
MDL Number MFCD00011382
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents