Please login to your online account to display your discounted pricing

Potassium Carbonate Anhydrous (Granular Powder/Certified ACS), Fisher Chemical


Quantity 250 lb.
Packaging Fiber Drum

Manufacturer: fisher scientific company  FLP208250LB

 View more versions of this product

Catalog No. P208250LB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 138.21
Melting Point 891°C
Quantity 250 lb.
Molecular Formula CK2O3
Color White
pH 11.5 to 12
Iron (Fe) 5ppm max.
Sodium (Na) 0.02% max.
Synonym Carbonic Acid Potassium Salt
Heavy Metals (as Pb) 5ppm max.
Packaging Fiber Drum
Assay Percent Range ≥99.0 %
Chemical Name or Material Potassium Carbonate Anhydrous
Identification Pass Test
MDL Number MFCD00011382
CAS 584-08-7
ChemAlert Storage Symbol Gray
Magnesium (Mg) 0.002% max.
Calcium (Ca) 0.005% max.
Insoluble Matter 0.01% max.
Phosphate 0.001% max.
Silica 0.005% max.
Sulfur Compounds 0.004% max.
Chloride 0.003% max.

Specifications

Chemical Name or Material Potassium Carbonate Anhydrous
CAS 584-08-7
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula CK2O3
Formula Weight 138.21
MDL Number MFCD00011382
Synonym Carbonic Acid Potassium Salt
Accessories
 

SDS & Safety Documents