Please login to your online account to display your discounted pricing

Potassium carbonate, ACS, 99.0% min, Alfa Aesar™

$121.92 - $121.92

Chemical Identifiers

Chemical Name or Material Potassium carbonate
CAS 584-08-7
Purity Grade ACS Grade
Molecular Formula K2CO3
Formula Weight 138.21
MDL Number MFCD00011382
Beilstein 4267587
Merck Index 14,7619
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA12609A1

  SDS
alfa aesar
12609A1
Each for $121.92
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 138.21
Quantity 1kg
Chemical Name or Material Potassium carbonate
TSCA TSCA
Physical Form Granular
Assay ≥99.0%
Sensitivity Hygroscopic
Molecular Formula K2CO3
MDL Number MFCD00011382
CAS 584-08-7
Beilstein 4267587
Merck Index 14,7619
Melting Point 891°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.29
RTECS Number TS7750000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.