Learn More
Potassium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012609A7
Description
It may be used in pharmaceutical laboratories as a drying agent or as a source of potassium. It is also used in fire extinguishers, to make soap, glass and to soften water. Aqueous potassium carbonate plays an important role in carbon dioxide absorption promoted by piperazine. It is also essential for the absorption of carbon dioxide promoted by boric acid. It is also useful for the synthesis of azoles and diazines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium carbonate | |
584-08-7 | |
Granules | |
99.0% min. | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
dipotassium carbonate | |
11430 | |
ACS | |
2.29 g/cm3 at 20°C |
891°C | |
Yellow | |
5 kg | |
CK2O3 | |
4267587 | |
Soluble in water. Insoluble in ethanol. | |
[K+].[K+].[O-]C([O-])=O | |
138.21 | |
138.21 | |
Hygroscopic |
Chemical Identifiers
584-08-7 | |
138.21 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
11430 |
CK2O3 | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-529-3
RTECSNumber : TS7750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only