missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium carbonate, ACS, 99.0% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1260914

 View more versions of this product

Catalog No. AA1260914



Specifications

Specifications

Potassium carbonate
99.0% min.
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium;carbonate
11430
Granular
2.29g/cm3 at 20°C
25g
584-08-7
CK2O3
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
C(=O)([O-])[O-].[K+].[K+]
138.205
138.21
ACS
Yellow

Chemical Identifiers

584-08-7
138.205
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
C(=O)([O-])[O-].[K+].[K+]
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
dipotassium;carbonate
Safety and Handling

Safety and Handling

H302-H315-H319-H335: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 209-529-3

Documents