Learn More
Potassium carbonate, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 198040010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent for analysisSpecifications
Potassium carbonate | |
Anhydrous | |
White | |
1 kg | |
CK2O3 | |
MFCD00011382 | |
15, 7740 | |
Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone | |
[K+].[K+].[O-]C([O-])=O | |
138.21 | |
138.21 | |
Analytical | |
Plastic bottle |
891.0°C | |
584-08-7 | |
Powder or Granular Powder | |
99+% | |
K2CO3 | |
01,1014; 04,50; 05,552; 08,408; 09,382; 10,323; 11,433; 12,403; 13,233; 15,167 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
dipotassium carbonate | |
11430 | |
≥99% | |
1% max. (300°C) |
Chemical Identifiers
584-08-7 | |
138.21 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
11430 |
CK2O3 | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Warning
RUO – Research Use Only