missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium carbonate, Puratronic™, 99.997% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1083814

 View more versions of this product

Catalog No. AA1083814



Specifications

Specifications

Potassium carbonate
99.99% (metals basis)
CK2O3
4267587
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
C(=O)([O-])[O-].[K+].[K+]
138.205
138.21
99.997%
2.29
584-08-7
(metals basis)
MFCD00011382
14,7619
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium;carbonate
11430
Powder
Puratronic™
25g

Chemical Identifiers

584-08-7
138.205
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
C(=O)([O-])[O-].[K+].[K+]
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
dipotassium;carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a

Warning

RTECSNumber : TS7750000

TSCA : TSCA

Documents