Please login to your online account to display your discounted pricing

Potassium bromomethyltrifluoroborate, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC431530050

 View more versions of this product

Catalog No. AC431530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 200.83
Quantity 5g
Color White to off-white
Infrared Spectrum Authentic
Molecular Formula CH2BBrF3K
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Potassium bromomethyltrifluoroborate
Assay Percent Range 95%
CAS 888711-44-2
Health Hazard 1 Warning
Elemental Analysis 5.62 to 6.33% C, 0.94 to 1.06%

Specifications

Chemical Name or Material Potassium bromomethyltrifluoroborate
Assay Percent Range 95%
Molecular Formula CH2BBrF3K
Formula Weight 200.83
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents