missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bromide, ultra dry, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1470014

 View more versions of this product

Catalog No. AA1470014



Description

Description

  • Ampouled under argon
Specifications

Specifications

Potassium bromide
7758-02-3
BrK
14,7618
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119.01
99.9%
−10 Mesh
Ampule
Hygroscopic
White
25g
Ultra dry
(metals basis)
MFCD00011358
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
Beads
2.75g/mL
Odorless
1.559
1435°C
730°C

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
BrK
MFCD00011358
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TS7650000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents