Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bromide, ultra dry, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Potassium bromide
CAS 7758-02-3
Name Note Ultra dry
Formula Weight 119.01
MDL Number MFCD00011358
Merck Index 14,7618
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1470014

 
alfa aesar™
1470014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1470022

 
alfa aesar™
1470022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1470036

 
alfa aesar™
1470036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 119.01
Packaging Ampule
Chemical Name or Material Potassium bromide
TSCA TSCA
Mesh Size −10 Mesh
Odor Odorless
Name Note Ultra dry
Physical Form Beads
Refractive Index 1.559
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00011358
CAS 7758-02-3
Merck Index 14,7618
Boiling Point 1435°C
Color White
Melting Point 730°C
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 2.75g/mL
RTECS Number TS7650000

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon