Learn More
Potassium bromide, Spectroscopy Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 039796Q4
Description
Manufacturing of photographic papers, process engraving and lithography, spectroscopy, as an analytical lab reagent. Potassium bromide is applied as a veterinary drug and antiepileptic medication for dogs. It is an utilitarian for the bromination of alkenes using cerium(IV) ammonium nitrate. It acts as an essential role with diacetoxy iodobenzene for the efficient
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1435°C | |
730°C | |
White | |
0.25 lb. | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119.01 | |
Odorless | |
Hygroscopic |
Potassium bromide | |
7758-02-3 | |
Random Cuttings, Chunks | |
BrK | |
14,7618 | |
Soluble in water and very slightly soluble in diethyl ether. | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
Spectroscopy | |
1.559 | |
2.75 g/mL |
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
potassium bromide |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 | |
[K+].[Br-] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
EINECSNumber : 231-830-3
RTECSNumber : TS7650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only