Please login to your online account to display your discounted pricing

Potassium Bromide Solution, For Mercury, LabChem™

$67.83 - $67.83

Chemical Identifiers

Chemical Name or Material Potassium Bromide Solution
CAS 7732-18-5
CAS Max % 72.28
CAS 7758-02-3
CAS Max % 27.72
Name Note For Mercury
Linear Formula KBr
Formula Weight 119
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187851

 
lab chem
LC187851
500mL Each for $67.83
Description & Specifications

Specifications

Formula Weight 119
Chemical Name or Material Potassium Bromide Solution
Viscosity 0.76cSt
Decomposition Information Hydrogen bromide
Color Clear, colorless
Quantity 500mL
Name Note For Mercury
Physical Form Liquid
Linear Formula KBr
Density 1.23g/mL
CAS 7732-18-5
CAS Max % 72.28
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 7758-02-3
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 27.72
Solubility Soluble in water
Specific Gravity 1.23g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.