missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bromide, Premium FTIR Grade

Manufacturer:  Alfa Aesar™ 47111KN

 View more versions of this product

Catalog No. AA47111KN



Specifications

Specifications

Potassium bromide
Premium FTIR
KBr
14,7618
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119.01
2.75g/mL
0.5g packets
1435°C
100 pc.
7758-02-3
BrK
MFCD00011358
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
Powder
Odorless
1.559
730°C

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
BrK
MFCD00011358
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TS7650000

TSCA : TSCA

SDS Documents