Learn More
Potassium bromide, FTIR Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03979414
Description
Potassium bromide is used as a veterinary drug, as an antiepileptic medication for dogs. It used for removal of peripheral membrane proteins. Potassium bromide and sodium perborate play an important role for mild and regioselective oxidative bromination of anilines. Its Pellet is useful for fine structural characteristics by infared spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1435°C | |
730°C | |
7758-02-3 | |
25 g | |
MFCD00011358 | |
Soluble in water and slightly soluble in diethyl ether. Insoluble in acetate. | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
Premium FTIR | |
Hygroscopic |
Potassium Bromide | |
FTIR Grade | |
Powder | |
BrK | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119.01 | |
1.559 | |
2.75 g/mL |
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
[K+].[Br-] |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
EINECSNumber : 231-830-3
RTECSNumber : TS7650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only