Please login to your online account to display your discounted pricing

Potassium bromide, EMD Millipore

For IR spectroscopy Uvasol™

$575.06 - $1,184.31

Chemical Identifiers

Formula Weight 119
Boiling Point 1435°C
CAS 7758-02-3
Chemical Name or Material Potassium bromide
Density 2.75g/cm3 at 20°C
Molecular Formula KBr
Solubility Information Soluble in water: 650g/L at 10°C
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry.
Synonym Hydrobromic Acid Potassium Salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1049070100

 
emd millipore corporation
1049070100*
Glass Bottle 100g Each for $575.06

M1049070500

 
emd millipore corporation
1049070500
Glass Bottle 500g Each for $1,184.31
Description & Specifications

Specifications

Formula Weight 119
Boiling Point 1435°C
pH 5.5 - 8.5 (50 g/l, H2O, 20°C)
Health Hazard 1 Irritant
Absorbance ≤ 0.015 (2g pressed disk, 0.5 cm layer thickness) (other ; 420-4000 cm⁻¹)
Quantity 100g
Melting Point 730°C
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry.
Molecular Formula KBr
Synonym Hydrobromic Acid Potassium Salt
Vapor Pressure 1.3hPa at 795°C
Odor Odorless
CAS 7758-02-3
Chemical Name or Material Potassium bromide
Solubility Information Soluble in water: 650g/L at 10°C
Density 2.75g/cm3 at 20°C
Packaging Glass Bottle
RTECS Number TS7650000
EINECS Number 231-830-3

Potassium bromide is transparent from the near UV to long wave IR wavelengths

Safety and Handling

Irritant

Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry.