missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromide, MilliporeSigma™

For IR spectroscopy Uvasol™

Manufacturer:  MilliporeSigma 1049070500

 View more versions of this product

Catalog No. M1049070500



Description

Description

Potassium bromide is transparent from the near UV to long wave IR wavelengths
Specifications

Specifications

Potassium bromide
BrK
Soluble in water: 650g/L at 10°C
[K+].[Br-]
119.00
CHEBI:32030
2.75g/cm3 at 20°C
Glass Bottle
1435°C
7758-02-3
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119
Odorless
1.3hPa at 795°C
500g

Chemical Identifiers

7758-02-3
119.00
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]
BrK
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

EINECSNumber : 231-830-3

RTECSNumber : TS7650000

Recommended Storage : Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry.

Documents
Promotions

Promotions