Please login to your online account to display your discounted pricing

Potassium Bromide, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™

$188.86 - $188.86

Chemical Identifiers

Chemical Name or Material Potassium Bromide
CAS 7732-18-5
CAS Max % 90.64
CAS 7758-02-3
CAS Max % 9.36
Purity Grade Certified Grade
Linear Formula KBr
Formula Weight 119
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187824

 
lab chem
LC187824
4L Each for $188.86
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 10.0% ± 0.5% (w/v)
Formula Weight 119
Chemical Name or Material Potassium Bromide
Storage Information Room Temperature
Physical Form Liquid
Solubility Soluble in water
Density 1.07g/mL
CAS Max % 90.64
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Specific Gravity 1.07g/mL
Identification Passes Test
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Linear Formula KBr
CAS 7732-18-5
Decomposition Information Hydrogen bromide
CAS Max % 9.36
CAS 7758-02-3
Quantity 4L
Traceability to NIST Traceable to NIST
Packaging Poly Bottle
Viscosity 0.9cSt
Color Clear, colorless
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.