missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Bromide (Beads or Powder/Certified ACS), Fisher Chemical

Manufacturer:  Thermo Scientific P2053

 View more versions of this product

Catalog No. P205-3




Specifications

Specifications

Potassium Bromide
Vapor Pressure: <0.01mmHg at 20°C
≥99%
BrK
IOLCXVTUBQKXJR-UHFFFAOYSA-M
[K+].[Br-]
119.00
CHEBI:32030
0.001% max.
0.002% max.
5ppm max.
0.005% max.
0.001% max.
0.001% max.
0.02% max.
<0.01 mmHg at 20°C
1435°C
734°C
3kg
7758-02-3
Certified ACS Grade
≥99 %
MFCD00011358
0.002% max.
potassium;bromide
253877
119
Solid
0.2% max.
Pass Test
0.001% max.
5ppm max.
Poly Bottle
0.005% max.
>800°C
White
5 to 8.8

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
CHEBI:32030
BrK
MFCD00011358
253877
Safety and Handling

Safety and Handling

SDS Documents