Please login to your online account to display your discounted pricing

Potassium Bromide, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™


Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Quantity 25g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  296103

 View more versions of this product

Catalog No. 02003719

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 118.99
CAS 7758-02-3
Molecular Formula KBr
Specific Gravity 1L = 2.75kg
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Quantity 25g
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Potassium Bromide

Specifications

Chemical Name or Material Potassium Bromide
CAS 7758-02-3
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula KBr
Formula Weight 118.99

    For Infrared Analysis