Please login to your online account to display your discounted pricing

Potassium Bromide, ACS Grade, ≥ 99.0%, LabChem™

$170.11 - $170.11

Chemical Identifiers

Chemical Name or Material Potassium Bromide
CAS 7758-02-3
CAS Max % 100
Purity Grade ACS Grade
Linear Formula KBr
Formula Weight 119
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187811

 
lab chem
LC187811
500g Each for $170.11
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥99.0%
Formula Weight 119
Chemical Name or Material Potassium Bromide
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Solubility Soluble in water
Specific Gravity 275g/cm3
Storage Information Room Temperature
Linear Formula KBr
CAS 7758-02-3
Decomposition Information Hydrogen bromide
Boiling Point 1,435°C
Color White
Melting Point 730°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of water and soap.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2,75g/cm3
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of water and soap.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.