missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromide, ACS, 99% min., Alfa Aesar™

For Manufacturing of Photographic Papers, Process Engraving and Lithography, Spectroscopy

Manufacturer:  Alfa Aesar 1261230

 View more versions of this product

Catalog No. AA1261230



Specifications

Specifications

Potassium bromide
7758-02-3
BrK
White powder/crystalline/beads
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119.01
ACS
White
For analysis ACS
99% min.
MFCD00011358
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
Crystalline
Odorless
250g

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
BrK
MFCD00011358
potassium bromide, potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

EINECSNumber : 231-830-3

Documents