Please login to your online account to display your discounted pricing

Potassium bromide, ACS, 99% min, Alfa Aesar™

Manufacturing of photographic papers, process engraving and lithography, spectroscopy, as an analytical lab reagent

Chemical Identifiers

Chemical Name or Material Potassium bromide
CAS 7758-02-3
Purity Grade ACS Grade
Molecular Formula KBr
Formula Weight 119.01
MDL Number MFCD00011358
Merck Index 14,7618
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA12612A7

  SDS
alfa aesar
12612A7
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 119.01
Quantity 5kg
Sensitivity Hygroscopic
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Health Hazard 1 Warning
MDL Number MFCD00011358
Solubility Information Freely soluble in water. Soluble in alcohol, glycerol
Color White
Density 2.75
Merck Index 14,7618
Health Hazard 2 GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Physical Form Crystalline
Molecular Formula KBr
Melting Point 730°C
Refractive Index 1.559
RTECS Number TS7650000
Chemical Name or Material Potassium bromide
CAS 7758-02-3
Assay ≥99%
TSCA TSCA
Boiling Point 1435°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.