Learn More
Potassium bromide, ACS, 99% min, Thermo Scientific Chemicals
For Manufacturing of Photographic Papers, Process Engraving and Lithography, Spectroscopy
Supplier: Thermo Scientific Chemicals 01261218
Description
Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1435°C | |
730°C | |
7758-02-3 | |
Crystalline | |
99% min. | |
MFCD00011358 | |
Freely soluble in water. Soluble in alcohol,glycerol. | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
ACS | |
1.559 | |
White powder/crystalline/beads |
Potassium bromide | |
For analysis ACS | |
White | |
50 g | |
BrK | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119.01 | |
Odorless | |
Hygroscopic | |
2.75 g/cm3 at 20°C |
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
[K+].[Br-] |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
EINECSNumber : 231-830-3
RTECSNumber : TS7650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only