Learn More
Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 222555000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
1435.0°C | |
734.0°C | |
7758-02-3 | |
5.0 to 8.8 (5% soln. at 20°C) | |
500 g | |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119 | |
Analytical | |
Plastic bottle |
Potassium bromide | |
p.a. | |
White | |
Crystalline Powder, Crystals and/or Chunks | |
99% min. (Argentometry) | |
KBr | |
15, 7739 | |
Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
≥99% | |
0.2% max. (130°C) |
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
[K+].[Br-] |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 231-830-3
RTECSNumber : TS7650000
TSCA : TSCA
RUO – Research Use Only