missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromide, 99.999%, (trace metal basis), Acros Organics™

Manufacturer:  Thermo Fisher Scientific AC436950500

 View more versions of this product

Catalog No. AC436950500



Description

Description

For ultra trace analysis
Specifications

Specifications

Potassium bromide
33% (K) Typical (Argentometry)
KBr
bromide salt of potassium
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119
99.999%
Plastic bottle
10ppm max.
White
50g
7758-02-3
BrK
15,7739
Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol
[K+].[Br-]
119
CHEBI:32030
Crystalline Powder
Trace Metal Basis
Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
1435.0°C
734.0°C

Chemical Identifiers

7758-02-3
119
bromide salt of potassium
CHEBI:32030
[K+].[Br-]
BrK
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 231-830-3

SDS Documents