missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromide, 99.92% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1098222

 View more versions of this product

Catalog No. AA1098222



Specifications

Specifications

Potassium bromide
99.92% (metals basis)
MFCD00011358
bromide salt of potassium
[K+].[Br-]
119.00
CHEBI:32030
Crystalline
Odorless
100g
7758-02-3
BrK
White Powder/crystalline/beads
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119.01
2.75g/cm3 at 20°C
Colorless

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
BrK
MFCD00011358
bromide salt of potassium
CHEBI:32030
[K+].[Br-]
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

EINECSNumber : 231-830-3

Documents