Please login to your online account to display your discounted pricing

Potassium Bromide, 40g + 100mL Water, Ricca Chemical


Quantity 1L
Packaging Poly natural

Manufacturer: ricca chemical company  RIR58800001A

 View more versions of this product

Catalog No. R58800001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Potassium Bromide
Name Note 40g + 100mL Water
CAS 7732-18-5
CAS 7758-02-3
Physical Form Liquid
Odor Odorless
Packaging Poly natural
Solvent or Matrix Aqueous Solution
Color Clear colorless
Quantity 1L

Specifications

Chemical Name or Material Potassium Bromide
Name Note 40g + 100mL Water
CAS 7732-18-5
CAS 7758-02-3
Physical Form Liquid